(±)-Camphorquinone 99%
Glass bottle, 50 g
: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
- :10373-78-1
- :C₁₀H₁₄O₂
- :MFCD00064160
- :166.22 g/mol
- :198…202 °C
- :Refrigerator
- :233-814-1
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:199 to 204 °C
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:50 g



