DI DELLADET VS2 2X5L I 1 * 2 Can
Catalog # DIVY7513526
Supplier: Diversey
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
pH tester, pH Checker®
Supplier: Hanna
VWR® Reagent Reservoirs
Supplier: VWR Collection
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Combitips® advanced PCR Clean - Positive Displacement Pipette Tips
Supplier: EPPENDORF