Order Entry
Denmark
Orders LinkContactUsLinkComponent
Total Organic Carbon (TOC) cleaning validation sets
Total Organic Carbon (TOC) cleaning validation sets
Low TOC vials (72) with blank labels, Clear, 40 ml
Catalog # 115-2152
Supplier:   Texwipe
Total Organic Carbon (TOC) cleaning validation sets
Catalog # 115-2152
Supplier:  Texwipe
Total Organic Carbon (TOC) cleaning validation sets
Total Organic Carbon (TOC) cleaning validation sets
Low TOC vials (72) with blank labels, Clear, 40 ml
Price
3.740,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
These sets contain components for sampling different locations in the production area, and provide an efficient means of transporting the samples from production to laboratory with minimal chance of contamination.
  • Low particle generation breakaway handle allows swab head to be placed into the vial with minimal handling and contamination
  • Good absorbent capacity, excellent chemical resistance
  • Autoclavable to ensure sampling under sterile conditions
Designed to simplify sampling for cleaning validation in the pharmaceutical industry.
Certifications: Certificates of analysis ensure traceability of the production lots.
Ordering information: Vials are certified to <10 ppb/l TOC. Swabs are certified <50 µg/l TOC.
Delivery information: Supplied with labels for identifying samples.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.
Specifications
  • Description:
    Low TOC vials (72) with blank labels
  • Seq:
    6
  • Vial capacity:
    40 ml
  • Cap colour:
    Clear