Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DI ACIPLUSFOAM VF59 20L W1779 1 * 20 L
Catalog # DIVY100844772
Supplier:
Diversey
Expand image
DI ACIPLUSFOAM VF59 20L W1779 1 * 20 L
Catalog # DIVY100844772
Supplier:
Diversey
Frequently Bought Together
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Labels, Hazardous Substance, GHS Labels
Supplier: VWR Collection
VWR® Nitrile Green Nitrile gloves
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection