Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

VWR®, COD calibration standards

COD standard solution, 600 mg/L, Plastic bottle, 500 mL
Katalog # 85066.260
Leverandør:   VWR Chemicals
VWR®, COD calibration standards
Katalog # 85066.260
Leverandør:  VWR Chemicals
CAS nummer:  
VWR®, COD calibration standards
VWR®, COD calibration standards

COD standard solution, 600 mg/L, Plastic bottle, 500 mL

CAS nummer:  
Price
1.160,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

These standards are ideal to use as control standards for COD measurements to verify that correct analysis for COD has taken place.

Achieving an acceptable result for a control standard will improve confidence in sample readings for this important environmental parameter.

Specifications

Specifications

  • Koncentration:
    600 mg/L
  • Pakning:
    Plastic bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    COD standard solution

Specifications