VWR®, COD calibration standards
COD standard solution, 30000 mg/L, Plastic bottle, 500 mL
These standards are ideal to use as control standards for COD measurements to verify that correct analysis for COD has taken place.
Achieving an acceptable result for a control standard will improve confidence in sample readings for this important environmental parameter.
Documents
- Concentration:30000 mg/L
- Pack type:Plastic bottle
- Pk:500 mL
- Description:COD standard solution