VWR®, COD calibration standards
COD standard solution, 1300 mg/L, Plastic bottle, 500 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
These standards are ideal to use as control standards for COD measurements to verify that correct analysis for COD has taken place.
Achieving an acceptable result for a control standard will improve confidence in sample readings for this important environmental parameter.
Documents
Certificates
Search by Lot NumberSDS_EN_CH_VWRC85054 SDS_EN_DE_VWRC85054 SDS_EN_PL_VWRC85061 SDS_EN_PL_VWRC85061.pdf SDS_EN_UK_VWRC85051.pdf
SDS
More
VWR® for Calibrations
Product Literature
Specifications
Specifications
- Concentration:1300 mg/L
- Pack type:Plastic bottle
- Pk:500 mL
- Description:COD standard solution