Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Bottles, narrow or wide neck, square or rectangular, with thread

50 ml, 25 mm, 36,1×42,7×63,5 mm
Catalog # RIXI2-D0-0050-25-K
Bottles, narrow or wide neck, square or rectangular, with thread
Catalog # RIXI2-D0-0050-25-K
Bottles, narrow or wide neck, square or rectangular, with thread
Bottles, narrow or wide neck, square or rectangular, with thread

50 ml, 25 mm, 36,1×42,7×63,5 mm

Price
3.320,00 kr
Log in for availability
1 * 1.400 Items
Product Details & Documents

HDPE, natural.

  • Rigid and shatter-proof
  • Resistant to water, aqueous solutions and most chemicals
  • Excellent water vapour resistance
  • Ribbed sides for secure grip

Square or rectangular bottles with large filling opening for packaging and storage of liquids, powders and granulates.

 : Screw caps must be ordered seperately.

Specifications
  • Capacity:
    50 ml
  • Seq:
    08
  • W×D×H:
    36,1×42,7×63,5 mm
  • Neck I-Ø:
    25 mm