Order Entry
Denmark
ContactUsLinkComponent
VWR® Low Retention Aerosol Filter Pipet tips. 1000uL Extra Long. Fit most 1000ul pipettors. RNase/DNase, Endotoxin, Protease, ATP Free, BPA Free. In hinged 96 tip racks. Racks are refillable. Clear polypropylene tips are autoclavable. 576 Tips per pa 1 * 1 SET
Catalog # VWRI732-1491BOM
Supplier:  VWR Collection
VWR® Low Retention Aerosol Filter Pipet tips. 1000uL Extra Long. Fit most 1000ul pipettors. RNase/DNase, Endotoxin, Protease, ATP Free, BPA Free. In hinged 96 tip racks. Racks are refillable. Clear polypropylene tips are autoclavable. 576 Tips per pa 1 * 1 SET
Catalog # VWRI732-1491BOM
Supplier:  VWR Collection
Frequently Bought Together