Order Entry
Denmark
ContactUsLinkComponent
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, K8 connector head, Diaphragm: HP Coatramic, Material: glass shaft, Electrolyte: Phermlyte, Length: 225 mm, Diameter: 12 mm, Process temperature: 0 to 140 C, Pr 1 * 1 items
Catalog # HAMI238626
Supplier:  HAMILTON BONADUZ
pH sensor with pre-pressurized electrolyte, Cleaning: SIP, CIP, Autoclavation, no cable, K8 connector head, Diaphragm: HP Coatramic, Material: glass shaft, Electrolyte: Phermlyte, Length: 225 mm, Diameter: 12 mm, Process temperature: 0 to 140 C, Pr 1 * 1 items
Catalog # HAMI238626
Supplier:  HAMILTON BONADUZ
Frequently Bought Together