Order Entry
Denmark
Orders LinkContactUsLinkComponent
Passive air sampling kits

Passive air sampling kits

Replacement tees with 0,0635 mm orifice, 30 ml/min, Siltek® treated stainless steel
Catalog # REST26339
Supplier:   Restek
Passive air sampling kits
Catalog # REST26339
Supplier:  Restek
Passive air sampling kits
Passive air sampling kits

Replacement tees with 0,0635 mm orifice, 30 ml/min, Siltek® treated stainless steel

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

This unique grab sampler minimises connections and leaks and houses a critical orifice in the tee fitting.

  • Different orifice sizes for sampling from four minutes to ten hours
  • Compression sample inlet allows easy connection
  • Canister valve connected to a sample port tubing
  • Stainless steel tee with orifice and vaccum gauge
  • Frit filter for insertion into a compression sample inlet

Specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.

 : Canisters and replacement tees and orifices can be ordered separately.

Specifications
  • Description:
    Replacement tees with 0,0635 mm orifice
  • Material:
    Siltek® treated stainless steel
  • Seq:
    904
  • Airflow:
    30 ml/min