Passive air sampling kits
Air sampler with 0,635 mm orifice, 30 ml/min, Siltek® treated stainless steel
Catalog # REST26337
Supplier: Restek
Jump to:
Product Details & Documents
This unique grab sampler minimises connections and leaks and houses a critical orifice in the tee fitting.
- Different orifice sizes for sampling from four minutes to ten hours
- Compression sample inlet allows easy connection
- Canister valve connected to a sample port tubing
- Stainless steel tee with orifice and vaccum gauge
- Frit filter for insertion into a compression sample inlet
Specifically designed for soil gas sampling by allowing the connection of tubing coming from the soil gas sample port.
Ordering information: Canisters and replacement tees and orifices can be ordered separately.
Specifications
- Description:Air sampler with 0,635 mm orifice
- Material:Siltek® treated stainless steel
- Seq:02
- Airflow:30 ml/min
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



