Packed Column 10% TCEP 100/120 Chromosorb PAW 2.5m, 2mm ID, Silco, Shimadzu 8A 1 * 1 items
REST80126-851
: Restek
Packed Column 10% TCEP 100/120 Chromosorb PAW 2.5m, 2mm ID, Silco, Shimadzu 8A 1 * 1 items
REST80126-851
: Restek
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK