Volumetric flasks, class A
250 ml, 14/15, 0,15 ± ml
Catalog # POULRB258/I/2
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass, with PP NS-stopper.
Individually numbered.
Certifications: DIN/BS/ISO 1042
Documents
POULRB258^WAC^J^2_TDS_EN
Datasheet
Specifications
- NS:14/15
- Capacity:250 ml
- Seq:26
- Tolerance:0,15 ± ml