Order Entry
Denmark
Orders LinkContactUsLinkComponent
Drum spill kits
Drum spill kits
Lab pack, maintenance, SKA-20
Katalog # BRDY813870
Leverandør:   Brady
Drum spill kits
Katalog # BRDY813870
Leverandør:  Brady
Drum spill kits
Drum spill kits
Lab pack, maintenance, SKA-20
Price
2.940,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
The drum spill kit is designed for medium sized spills.
  • Water and chemical resistant drum protects sorbents
  • Protects sorbents against moisture, dirt and damage
  • Drum can also be used as shipping container for used sorbents
  • Easy access to products with screw top lid
Delivery information: Supplied with 12 pads (410×510 mm), 3 socs (Ø 7,6×3660 mm), 2 pillows (430×480 mm), 1 pair of gloves, 1 pair of goggles and 3 disposal bags.
Specifikationer
  • Description:
    Lab pack, maintenance, SKA-20
  • Seq:
    02