Multi layer adhesive floor mats
Blue, 660×1143 mm, 0,036 mm
Catalog # PURUPM264538B
Supplier: PURUS
Jump to:
Product Details & Documents
A choice of either 30 or 60 layer adhesive entrance mats are manufactured from low density polyethylene. The mats control contamination by removing debris from the soles of shoes and the wheels of transport containers. When one layer is dirty, simply remove by the numbered corner tab, leaving a fresh layer ready for use.
- Antimicrobial agent protects against the growth of bacteria and fungi
- Non-slip, low profile, individually numbered layers
- Reduced maintenance and cleaning costs
Colour: Clear, white, grey or blue
Specifications
- Colour:Blue
- L×W:660×1143 mm
- Thickness:0,036 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



