PERCHLORIC ACID 0.1 MOL/L (0.1N) IN ACET 1 * 1.000 mL
Catalog # APPC181046.1611
Supplier: PanReac AppliChem
PERCHLORIC ACID 0.1 MOL/L (0.1N) IN ACET 1 * 1.000 mL
Catalog # APPC181046.1611
Supplier: PanReac AppliChem
Frequently Bought Together
Svovlsyre 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Ansigtsservietter
Supplier: VWR Collection
pH-teststrimler, MQuant®, Supelco®
Supplier: Merck
Kaliumchlorid 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
VWR®, Nitril handsker, NITRILE
Supplier: VWR Collection
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Glycerin 85% Ph. Eur.
Supplier: VWR Chemicals
Eddikesyre glacial 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Natriumthiosulfat standardised solution 0.1 N i vandigopløsning
Supplier: Thermo Fisher Scientific
Colecalciferol 99%
Supplier: Thermo Fisher Scientific
Ammoniumchlorid ≥98%
Supplier: Thermo Fisher Scientific