Nitric Acid 69% for analysis, ACS, ISO 1 * 1.000 mL
Catalog # APPC131037.1611
Supplier: PanReac AppliChem
Nitric Acid 69% for analysis, ACS, ISO 1 * 1.000 mL
Catalog # APPC131037.1611
Supplier: PanReac AppliChem
Frequently Bought Together
Cartridges for Arium®
Supplier: Sartorius
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Beakers, with handle, PP
Supplier: BURKLE
Sulphuric acid 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck