Hydrochloric Acid 37% for analysis, ACS, ISO 1 * 1.000 mL
APPC131020.1211
Frequently Bought Together
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
: MACRON AVANTOR BRAND
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
: Thermo Fisher Scientific
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
: MACRON AVANTOR BRAND
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
: Thermo Fisher Scientific
Petroleum spirit, 30…50 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
n-Pentane 99+%, extra pure
: Thermo Fisher Scientific
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals