G-Biosciences Immobilized Heparin is a ready-to-use purification resin for a wide range of proteins. The resin consists of 4% cross -linked agarose covalently coupled to heparin through amide bonds. 1 * 5 mL
Catalog # GENO786-842
Supplier: G-Biosciences
Frequently Bought Together