Order Entry
Denmark
ContactUsLinkComponent
G-Biosciences Silica Magnetic Beads are Fe3O4 magnetic beads coated with a silicon dioxide (SiO2) layer. Since silica is able to bin d to the nucleic acids, G-Biosciences Silica Magnetic Beads serve as a simple and efficient tool for plasmid DNA puri 1 * 100 mL
Catalog # GENO786-917
Supplier:  G-Biosciences
G-Biosciences Silica Magnetic Beads are Fe3O4 magnetic beads coated with a silicon dioxide (SiO2) layer. Since silica is able to bin d to the nucleic acids, G-Biosciences Silica Magnetic Beads serve as a simple and efficient tool for plasmid DNA puri 1 * 100 mL
Catalog # GENO786-917
Supplier:  G-Biosciences