The fixing of proteins in polyacrylamide gels prevents the diffusion of the proteins, thus keeping protein bands sharp and correctly resolved, and washes away running buffer components that may interfere with other downstream applications, such as s 1 * 1 L
Catalog # GENO786-236
Supplier: G-Biosciences
Frequently Bought Together