Order Entry
Denmark
ContactUsLinkComponent
G-Biosciences IgG Binding, Wash & Elution Buffers are designed for the non-denaturing, high yield purification of antibodies from Ig G affinity purification resins. The buffers are ideal for the purification of antibodies from Protein A, Protein G r 1 * 100 mL
Catalog # GENO786-200
Supplier:  G-Biosciences
G-Biosciences IgG Binding, Wash & Elution Buffers are designed for the non-denaturing, high yield purification of antibodies from Ig G affinity purification resins. The buffers are ideal for the purification of antibodies from Protein A, Protein G r 1 * 100 mL
Catalog # GENO786-200
Supplier:  G-Biosciences