G-Biosciences IgG Binding, Wash & Elution Buffers are designed for the non-denaturing, high yield purification of antibodies from Ig G affinity purification resins. The buffers are ideal for the purification of antibodies from Protein A, Protein G r 1 * 1 L
Catalog # GENO786-205
Supplier: G-Biosciences
Frequently Bought Together