Order Entry
Denmark
ContactUsLinkComponent
Glutathione Resin is designed for the affinity purification of proteins with a glutathione S-transferase (GST) tag. The resin consis ts of reduced glutathione (GSH) covalently coupled to 4% cross-linked agarose, via a 10-carbon spacer arm. The resin 1 * 100 µG
Catalog # GENO786-785
Supplier:  G-Biosciences
Glutathione Resin is designed for the affinity purification of proteins with a glutathione S-transferase (GST) tag. The resin consis ts of reduced glutathione (GSH) covalently coupled to 4% cross-linked agarose, via a 10-carbon spacer arm. The resin 1 * 100 µG
Catalog # GENO786-785
Supplier:  G-Biosciences
Frequently Bought Together