Order Entry
Denmark
ContactUsLinkComponent
The SpinOUT™ GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5µl to 4ml sample volumes. Spin-OUT™ GT-100 is for the purification o 1 * 5 mL
Catalog # GENO786-868
Supplier:  G-Biosciences
The SpinOUT™ GT-100, GT-600 and GT-1200 columns are versatile, spin-format columns for the desalting and buffer exchange of peptide, protein and nucleic acid solutions ranging from 5µl to 4ml sample volumes. Spin-OUT™ GT-100 is for the purification o 1 * 5 mL
Catalog # GENO786-868
Supplier:  G-Biosciences