Order Entry
Denmark
ContactUsLinkComponent
FLUORESCENT DYE 750-I PHALLOIDIN FLUORESCENCE DYE 750-I WITH PHALLOIDIN CONJUGATE. COMPARED WITH ALEXA FLUOR 750, FLUORESCENT DYE 7 50-I IS MUCH BRIGHTER WITH ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 750-I IS WELL SEPARATED FRO 1 * 300 Assays
Catalog # ABNOU0305
Supplier:  Abnova
FLUORESCENT DYE 750-I PHALLOIDIN FLUORESCENCE DYE 750-I WITH PHALLOIDIN CONJUGATE. COMPARED WITH ALEXA FLUOR 750, FLUORESCENT DYE 7 50-I IS MUCH BRIGHTER WITH ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 750-I IS WELL SEPARATED FRO 1 * 300 Assays
Catalog # ABNOU0305
Supplier:  Abnova