Order Entry
Denmark
ContactUsLinkComponent
FLUORESCENT DYE 750-I STREPTAVIDIN FLUORESCENCE DYE 750-I WITH STREPTAVIDIN CONJUGATE. COMPARED WITH ALEXA FLUOR 750, FLUORESCENT D YE 750-I IS MUCH BRIGHTER WITH ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 750-I IS WELL SEPARATED 1 * 200 µG
Catalog # ABNOU0304
Supplier:  Abnova
FLUORESCENT DYE 750-I STREPTAVIDIN FLUORESCENCE DYE 750-I WITH STREPTAVIDIN CONJUGATE. COMPARED WITH ALEXA FLUOR 750, FLUORESCENT D YE 750-I IS MUCH BRIGHTER WITH ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 750-I IS WELL SEPARATED 1 * 200 µG
Catalog # ABNOU0304
Supplier:  Abnova