Order Entry
Denmark
ContactUsLinkComponent
FLUORESCENT DYE 700-I STREPTAVIDIN FLUORESCENCE DYE 700-I WITH STREPTAVIDIN CONJUGATE. IN COMPARISON WITH ALEXA FLUOR 700, FLUORESC ENCE DYE 700-I ARE BRIGHTER WITH STRONGER ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 700-I IS WEL 1 * 200 µG
Catalog # ABNOU0301
Supplier:  Abnova
FLUORESCENT DYE 700-I STREPTAVIDIN FLUORESCENCE DYE 700-I WITH STREPTAVIDIN CONJUGATE. IN COMPARISON WITH ALEXA FLUOR 700, FLUORESC ENCE DYE 700-I ARE BRIGHTER WITH STRONGER ABSORPTION AT 633NM. FLUORESCENCE EMISSION OF FLUORESCENCE DYE 700-I IS WEL 1 * 200 µG
Catalog # ABNOU0301
Supplier:  Abnova