Order Entry
Denmark
ContactUsLinkComponent
IL31 (MOUSE) ELISA KIT IL31 (MOUSE) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF MOUSE IL31 1 Ki t 1 * 1 KIT
Catalog # ABNOKA4226
Supplier:  Abnova
IL31 (MOUSE) ELISA KIT IL31 (MOUSE) ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT OF MOUSE IL31 1 Ki t 1 * 1 KIT
Catalog # ABNOKA4226
Supplier:  Abnova
Frequently Bought Together