Double bossheads
With nickel plated brass screws, Zinc die-cast, nickel plated, 16,0 mm
Catalog # USBE2203
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
For stand assembly.
- Quick, easy assembly and disassembly
- Robust, durable
- Wide range of different materials available for many applications
Specifications
- Description:With nickel plated brass screws
- Material:Zinc die-cast, nickel plated
- Seq:6
- For rod Ø:16,0 mm