Tweezer for weights from 50g to 1mg, length 100mm, stainless steel anti-magnetic, tip protected by PVC. 1 * 1 items
ZWIEP6660000
: ZWIEBEL
Tweezer for weights from 50g to 1mg, length 100mm, stainless steel anti-magnetic, tip protected by PVC. 1 * 1 items
ZWIEP6660000
: ZWIEBEL
Frequently Bought Together
Accessories for calibration weights, E1, E2, F1, F2
: Sartorius Balances
VWR®, Digital Timer, One Channel
: VWR Collection
VWR®, Mini vortex mixer
: VWR Collection
VWR®, Digital Timer, One Channel
: VWR Collection
VWR® Pipette Controller
: VWR Collection
Accessories for MA series
: Sartorius Balances
Eppendorf® Pipette Holder System
: EPPENDORF
VWR®, Digital Timer, Four Channels, with Clock
: VWR Collection
Gas cartridges, Campingaz®
: CAMPING GAZ
Cartridge 220G for cartridge burner
: USBECK Laborgeräte
Vortex mixers, Vortex-Genie®
: SCIENTIFIC INDUSTRIES ELMIS
Cartridges for Milli-Q®
: Merck
VWR®, Calibration Weights, Class F2
: VWR Collection