Sample diluent buffer 1 * 30 mL
Catalog # BSBTAR1106-1
Supplier: Boster Bio
Frequently Bought Together
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1-Pentanol ≥98.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals