Order Entry
Denmark
ContactUsLinkComponent
A/D-WANDLER INTERFACE INKL 1 KABEL SET For usage of only one channel => SCSO35900-60900 => SCSO35900-60920 For usage of the second channel please order additional 1x SCSO35900-CABLESET 1 * 1 items
Catalog # SCSO35900E2SET
Supplier:  VWR Collection
A/D-WANDLER INTERFACE INKL 1 KABEL SET For usage of only one channel => SCSO35900-60900 => SCSO35900-60920 For usage of the second channel please order additional 1x SCSO35900-CABLESET 1 * 1 items
Catalog # SCSO35900E2SET
Supplier:  VWR Collection
Frequently Bought Together