PYRENE (13C3, 99%) 100 UG/ML 1 * 1,2 ml
Catalog # CAMBCLM-3601-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
o-Xylene ≥98.0% (by GC)
Supplier: TCI
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Toluene 99.5% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals