PHENANTHRENE (13C6, 99%) 100 UG/ML 1 * 1,2 ml
Catalog # CAMBCLM-2451-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
PHENANTHRENE (13C6, 99%) 100 UG/ML 1 * 1,2 ml
Catalog # CAMBCLM-2451-1.2
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile, Prepsolv® for preparative chromatography
Supplier: Merck
Ethanol absolute, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Acetone, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
n-Heptane
Supplier: Merck
Vanillin for synthesis, Sigma-Aldrich®
Supplier: Merck
Pyridine, EMPARTA® ACS analytical reagent
Supplier: Merck
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8% for peptide synthesis
Supplier: VWR Chemicals
o-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
PYRENE (13C3, 99%) 100 UG/ML 1 * 1,2 ml
Supplier: CAMBRIDGE ISOTOPE LABS