Digital dry block heaters
Dry Block Heater 3, 3, 330 W, RT*...120 °C, ±0,2 °C, 151×380×73 mm, 1,6 kg
460-0261
Digital block heaters which hold from one up to four aluminum blocks, provide precise temperature control in small vessels. The PT1000 temperature sensor allows temperature control directly in the sample vessel. It is used with PCR tubes, PCR strips, Greiner tubes, microplates and cuvettes. The block heaters are ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood/urea nitrogen determinations and in situ hybridisations.
- Stepless adjustable temperature from ambient up to 120 °C
- Timer: Countdown, adjustable from 1 min to 99h 59 min
- Counter: Shows heating time
- Highly versatile with a large number of aluminium alloy blocks
- Fixed safety circuit and error code display
- Acoustic signal at end of test
: Supplied with PT1000 temperature sensor, but without heating blocks, blocks must be ordered separately.
: * RT = Ambient
- Heating capacity:330 W
- No. of blocks:3
- Temperature Range:RT*...120 °C
- Weight:1,6 kg
- Seq:05
- W×D×H:151×380×73 mm
- Temp. stability:±0,2 °C
- Type:Dry Block Heater 3
Accessories
- DescriptionForDepth
- DescriptionTypeRange
- 460-1041Temperature sensor, coated (SafeCoat), DIN 45322PT 1000.90−10...+260 °C1 * 1 Items - 2.860,00 kr
- DescriptionForDepth
- 460-1036Single block, 80× 6,4 mm Ø stripsPCR tube strips (10×8 strips)15,5 mm1 * 1 Items - 1.120,00 kr
- DescriptionForDepth
- DescriptionForDepth
- 460-1025Single block, 4×35 mm Ø holesTubes, 35 mm, with round bottom47,6 mm1 * 1 Items - 1.180,00 kr
- DescriptionForDepth
- 460-1022Single block, 12×17,5 mm Ø holesTubes, 15 - 16 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- DescriptionForDepth
- DescriptionForDepth
- 460-1018Single block, 30×8,3 mm Ø holesTubes, 6 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- DescriptionForDepth
- DescriptionForDepth
- 460-1020Single block, 16×13,9 mm Ø holesTubes, 12 - 13 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- Description
- 442-2041Support rod, stainless steel, length 450 mm, Ø 10 mm, for all stirrers with M10 thread1 * 1 Items - 707,00 kr
- DescriptionForDepth
- DescriptionForDepth
- DescriptionForDepth
- DescriptionForDepth
- 460-1016Single block, combination: 4/3/2×1,5/15/50 mlCentrifuge tubes, 1,5 ml/15 ml/50 ml39,1/44,5/47,6 mm1 * 1 Items - 1.120,00 kr
- DescriptionForDepth
- DescriptionForDepth
- DescriptionForDepth
- DescriptionForDepth
- 460-1021Single block, 20×13,9 mm Ø holesTubes, 12 - 13 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- DescriptionForDepth
- 460-1019Single block, 24×10,7 mm Ø holesTubes, 10 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- DescriptionForDepth
- DescriptionForDepth
- 460-1011Single block, 20×11,5 mm Ø holesVWR/Eppendorf Tubes®, 2,0 ml38,1 mm1 * 1 Items - 1.120,00 kr
- DescriptionForDepth
- DescriptionForDepth
- 460-1023Single block, 8×21 mm Ø holesTubes, 20 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- Description
- DescriptionForDepth
- 460-1026Single block, 12×19,1 mm Ø holesTubes, 17 - 18 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- DescriptionForDepth
- 460-1017Single block, combination: 6/10/5×0,5/1,5/2 mlMicrotubes, 0,5 ml/1,5 ml/2,0 ml27,6/39,1/38,1 mm1 * 1 Items - 1.120,00 kr
- DescriptionForDepth
- DescriptionForDepth
- DescriptionForDepth
- DescriptionForDepth
- 460-1024Single block, 6×26,2 mm Ø holesTubes, 25 mm, with round bottom48,4 mm1 * 1 Items - 1.180,00 kr
- DescriptionForDepth
- Description
- DescriptionForDepth
- DescriptionForDepth
- 460-1015Single block, combination: 6/5/3×6/12/13/25 mm Ø holesTest tubes, 6 ml/12 - 13 ml/25 ml48,4/48,4/48,4 mm1 * 1 Items - 1.120,00 kr
- DescriptionForDepth
- DescriptionForDepth
- DescriptionTypeRange
- 442-0584Stainless steel temperature sensor, DIN 45322PT 1000.60−20...+350 °C1 * 1 Items - 1.350,00 kr