POTASSIUM HYDROXIDE ALCOHOLIC 2M 1 * 100 mL
Katalog # ACSD1070301
Leverandør: CPACHEM
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Leverandør: Merck
m-Xylene for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ethanol, GPR RECTAPUR®
Leverandør: VWR Chemicals
n-Heptane, anhydrous 99+%
Leverandør: Thermo Fisher Scientific
Hydrindantin dihydrate, Supelco®
Leverandør: Merck
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Leverandør: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99+% ACS
Leverandør: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Leverandør: Honeywell Chemicals
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Leverandør: PanReac AppliChem
Acetonitrile ≥99%, TECHNICAL
Leverandør: VWR Chemicals
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Leverandør: Thermo Fisher Scientific
Toluene 99.5% ACS for spectroscopy
Leverandør: Thermo Fisher Scientific
2-Propanol 1 * 500 mL
Leverandør: Wako
Chloroform 99.8% stabilised for spectroscopy
Leverandør: Thermo Fisher Scientific
Tetrahydrofuran ≥99.0% contains inhibitor, Reagent Grade
Leverandør: Honeywell Chemicals