Drape sheets, Foliodrape® Protect
Foliodrape® Protect drape sheets, sterile, in 4-layer packaging, 900×1500 mm
Catalog # HART277506/1
Supplier: HARTMANN PAUL
Jump to:
Product Details & Documents
These drape sheets are made from a soft polyethylene foil laminated to an absorbent non woven top layer. For sterile draping of patients and equipment during operations with moderate quantities of fluids, especially in outpatient settings.
- Impermeable to moisture and bacteria
- Abrasion resistant and extremely low linting
- Highly conformable
- Also available as an adhesive drape with self-adhesive area for rapid and accurate positioning at the incision site
Certifications: DIN 13795 (1-3)
Documents
Specifications
- Description:Foliodrape® Protect drape sheets, sterile, in 4-layer packaging
- L×W:900×1500 mm