Order Entry
Denmark
ContactUsLinkComponent
Cryotube Cryo.S 2ml polypropylene internal thread self standing natural round bottom natural screw cap steril
Catalog # GREI122261UMC
Supplier:  Greiner Bio-One
Cryotube Cryo.S 2ml polypropylene internal thread self standing natural round bottom natural screw cap steril
Catalog # GREI122261UMC
Supplier:  Greiner Bio-One