Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydrometers in plastic

Hydrometers in plastic

0 - 50 %mas/°B, 0,5 %mas/°B, 300 mm
Catalog # SCNL3002595
Supplier:   Ludwig Schneider
Hydrometers in plastic
Catalog # SCNL3002595
Supplier:  Ludwig Schneider
Hydrometers in plastic
Hydrometers in plastic

0 - 50 %mas/°B, 0,5 %mas/°B, 300 mm

Price
741,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

High quality polycarbonate hydrometers for %mas/°Brix measures.

  • Breakage protected (no glass breakage)
  • High transparency with easy-to-read scales
  • High precision and reproducibility

Suitable for applications in food, pharmaceutical and cosmetic industries.

Reference temperature: 20 °C

Ordering information: Available with works calibration certificate on request.

Packaging: In square plastic sleeve with individual serial number.

Specifications
  • Length:
    300 mm
  • Seq:
    08
  • Division:
    0,5 %mas/°B
  • Range:
    0 - 50 %mas/°B
  • Cat. No.:
    SCNL3002595
  • Reference temperature:
    20 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.