Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Hydrometers in plastic

30 - 45 %mas/°B, 0,2 %mas/°B, 250 mm
  SCNL3003045
Hydrometers in plastic
  SCNL3003045
Hydrometers in plastic
Hydrometers in plastic

30 - 45 %mas/°B, 0,2 %mas/°B, 250 mm

 
741,00 kr
   
1 * 1 Items
 

High quality polycarbonate hydrometers for %mas/°Brix measures.

  • Breakage protected (no glass breakage)
  • High transparency with easy-to-read scales
  • High precision and reproducibility

Suitable for applications in food, pharmaceutical and cosmetic industries.

Reference temperature: 20 °C

 : Available with works calibration certificate on request.

 : In square plastic sleeve with individual serial number.

 
  • Length:
    250 mm
  • Seq:
    03
  • Division:
    0,2 %mas/°B
  • Range:
    30 - 45 %mas/°B
  • Cat. No.:
    SCNL3003045
  • Reference temperature:
    20 °C