Order Entry
Denmark
ContactUsLinkComponent
Pack of 10 Mini data logger testo 184 T1 for temperature (operating time 90 days) All in one data logger: PDF measurement data, configuration file, calibration certificate and instruction manual. Immediate creation of a PDF report directly after con 1 * 10 items
Catalog # 620-2307
Supplier:  Testo
Pack of 10 Mini data logger testo 184 T1 for temperature (operating time 90 days) All in one data logger: PDF measurement data, configuration file, calibration certificate and instruction manual. Immediate creation of a PDF report directly after con 1 * 10 items
Catalog # 620-2307
Supplier:  Testo
Frequently Bought Together