Order Entry
Denmark
ContactUsLinkComponent
Rofecoxib is a non-steroidal anti-inflammatory drug that inhibits cyclooxygenase (COX)-2 activity with 1,000-fold selectivity over t hat of COX-1 (IC50s = 26 nM and > 50 µM in human osteosarcoma cells).{12018} In in vivo rodent models, rofecoxib can 1 * 50 mg
Catalog # CAYM10010260-50
Supplier:  Cayman Chemical
Rofecoxib is a non-steroidal anti-inflammatory drug that inhibits cyclooxygenase (COX)-2 activity with 1,000-fold selectivity over t hat of COX-1 (IC50s = 26 nM and > 50 µM in human osteosarcoma cells).{12018} In in vivo rodent models, rofecoxib can 1 * 50 mg
Catalog # CAYM10010260-50
Supplier:  Cayman Chemical
Frequently Bought Together