qPCR detection of Infectious Hematopoietic Necrosis Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only) , Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-IHNV
Supplier: PRIMERDESIGN
Frequently Bought Together