Order Entry
Denmark
ContactUsLinkComponent
qPCR detection of H1N1 influenza. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogen ous control, extraction control 1 * 150 Tests
Katalog # PATH-H1N1
Supplier:  PRIMERDESIGN
qPCR detection of H1N1 influenza. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogen ous control, extraction control 1 * 150 Tests
Katalog # PATH-H1N1
Supplier:  PRIMERDESIGN
Frequently Bought Together