Order Entry
Denmark
ContactUsLinkComponent
qPCR detection of Plasmodium malariae. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, En dogenous control, extraction control 1 * 150 Tests
Katalog # PATH-P.MALARIA
Supplier:  PRIMERDESIGN
qPCR detection of Plasmodium malariae. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, En dogenous control, extraction control 1 * 150 Tests
Katalog # PATH-P.MALARIA
Supplier:  PRIMERDESIGN
Frequently Bought Together