Order Entry
Denmark
ContactUsLinkComponent
qPCR detection of Respiratory Syncytial Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard c urve, Endogenous control, extraction control 1 * 150 Tests
  Z-PATH-RSV-SPP
qPCR detection of Respiratory Syncytial Virus. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard c urve, Endogenous control, extraction control 1 * 150 Tests
  Z-PATH-RSV-SPP
Frequently Bought Together