qPCR detection of Plasmodium knowlesi. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vir uses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-P.KNO
Supplier: PRIMERDESIGN
qPCR detection of Plasmodium knowlesi. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA genome vir uses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-P.KNO
Supplier: PRIMERDESIGN