Order Entry
Denmark
ContactUsLinkComponent
qPCR detection of Cyprinid herpesvirus 3. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
  PATH-CYHV-3
qPCR detection of Cyprinid herpesvirus 3. Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
  PATH-CYHV-3
Frequently Bought Together